Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methoxy-1-indanone, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
13623-25-1
C10H10O2
162.19
UJGDLLGKMWVCPT-UHFFFAOYSA-N
6-methoxy-1-indanone,6-methoxy-1h-indanone,6-methoxy-indan-1-one,6-methoxy-2,3-dihydro-1h-inden-1-one,6-methoxyindan-1-one,2,3-dihydro-6-methoxyinden-1-one,1h-inden-1-one, 2,3-dihydro-6-methoxy,6-methoxyindanone,6-methoxy indanone,6-methoxy-1-indanon
334036
6-methoxy-2,3-dihydroinden-1-one
COC1=CC2=C(CCC2=O)C=C1
13623-25-1
C10H10O2
162.19
UJGDLLGKMWVCPT-UHFFFAOYSA-N
6-methoxy-1-indanone,6-methoxy-1h-indanone,6-methoxy-indan-1-one,6-methoxy-2,3-dihydro-1h-inden-1-one,6-methoxyindan-1-one,2,3-dihydro-6-methoxyinden-1-one,1h-inden-1-one, 2,3-dihydro-6-methoxy,6-methoxyindanone,6-methoxy indanone,6-methoxy-1-indanon
334036
6-methoxy-2,3-dihydroinden-1-one
COC1=CC2=C(CCC2=O)C=C1
Quantity 1g
Packaging Glass bottle
Color White to Yellow
Melting Point 107.0°C to 111.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethyl acetate,methylene dichloride
Formula Weight 162.19
Physical Form Fine Crystalline Powder
Percent Purity 98%
Chemical Name or Material 6-Methoxy-1-indanone
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning