Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methoxy-1-tetralone, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
1078-19-9
C11H12O2
176.21
MFCD00001695
MNALUTYMBUBKNX-UHFFFAOYSA-N
6-methoxy-1-tetralone,6-methoxy-3,4-dihydronaphthalen-1 2h-one,6-methoxytetralone,6-methoxy-alpha-tetralone,1 2h-naphthalenone, 3,4-dihydro-6-methoxy,6-methoxy-1,2,3,4-tetrahydronaphthalen-1-one,6-methoxy-3,4-dihydro-1 2h-naphthalenone,6-methoxytetralin-1-one,3,4-dihydro-6-methoxy-1 2h-naphthalenone,6-methoxy-.alpha.-tetralone
14112
6-methoxy-3,4-dihydro-2H-naphthalen-1-one
COC1=CC2=C(C=C1)C(=O)CCC2
1078-19-9
C11H12O2
176.21
MFCD00001695
MNALUTYMBUBKNX-UHFFFAOYSA-N
6-methoxy-1-tetralone,6-methoxy-3,4-dihydronaphthalen-1 2h-one,6-methoxytetralone,6-methoxy-alpha-tetralone,1 2h-naphthalenone, 3,4-dihydro-6-methoxy,6-methoxy-1,2,3,4-tetrahydronaphthalen-1-one,6-methoxy-3,4-dihydro-1 2h-naphthalenone,6-methoxytetralin-1-one,3,4-dihydro-6-methoxy-1 2h-naphthalenone,6-methoxy-.alpha.-tetralone
14112
6-methoxy-3,4-dihydro-2H-naphthalen-1-one
COC1=CC2=C(C=C1)C(=O)CCC2
Boiling Point 171.0°C (11.0 mmHg)
Packaging Glass bottle
Melting Point 77.0°C to 79.0°C
Color Brown to Yellow
Quantity 100g
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 09, II, 889
Merck Index 15, 6074
Formula Weight 176.21
Physical Form Fine Crystalline Powder
Percent Purity 99%
Chemical Name or Material 6-Methoxy-1-tetralone
Show More Show Less