Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Methoxy-2-methylquinoline, 97%

Click to view available options
Quantity:
1g
5g
1078-28-0
C11H11NO
173.215
MFCD00006761
NAGJQQFMJKMXJQ-UHFFFAOYSA-N
6-methoxyquinaldine,2-methyl-6-methoxyquinoline,quinoline, 6-methoxy-2-methyl,quinaldine, 6-methoxy,6-methoxy-2-methyl-quinoline,quinoline,6-methoxy-2-methyl,pubchem5910,acmc-2098xp,2-methy-6-methoxyquinolinel
70648
6-methoxy-2-methylquinoline
CC1=NC2=C(C=C1)C=C(C=C2)OC
1078-28-0
C11H11NO
173.215
MFCD00006761
NAGJQQFMJKMXJQ-UHFFFAOYSA-N
6-methoxyquinaldine,2-methyl-6-methoxyquinoline,quinoline, 6-methoxy-2-methyl,quinaldine, 6-methoxy,6-methoxy-2-methyl-quinoline,quinoline,6-methoxy-2-methyl,pubchem5910,acmc-2098xp,2-methy-6-methoxyquinolinel
70648
6-methoxy-2-methylquinoline
CC1=NC2=C(C=C1)C=C(C=C2)OC
Quantity 5g
Flash Point 47°C (117°F)
Melting Point 62°C to 66°C
Formula Weight 173.21
Percent Purity 97%
Chemical Name or Material 6-Methoxy-2-methylquinoline

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning