Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Methoxy-2-naphthaldehyde, 99%

Click to view available options
Quantity:
5g
25g
100g
3453-33-6
C12H10O2
186.21
MFCD00081152
VZBLASFLFFMMCM-UHFFFAOYSA-N
6-methoxy-2-naphthaldehyde,6-methoxy-2-naphthalaldehyde,unii-9244l5l5zy,6-methoxy-naphthalene-2-carbaldehyde,2-naphthalenecarboxaldehyde, 6-methoxy,pubchem13704,acmc-1ah4r,6-methoxy-2-napthaldehyde,dsstox_cid_31510,dsstox_rid_97395
76991
6-methoxynaphthalene-2-carbaldehyde
COC1=CC2=C(C=C1)C=C(C=C2)C=O
3453-33-6
C12H10O2
186.21
MFCD00081152
VZBLASFLFFMMCM-UHFFFAOYSA-N
6-methoxy-2-naphthaldehyde,6-methoxy-2-naphthalaldehyde,unii-9244l5l5zy,6-methoxy-naphthalene-2-carbaldehyde,2-naphthalenecarboxaldehyde, 6-methoxy,pubchem13704,acmc-1ah4r,6-methoxy-2-napthaldehyde,dsstox_cid_31510,dsstox_rid_97395
76991
6-methoxynaphthalene-2-carbaldehyde
COC1=CC2=C(C=C1)C=C(C=C2)C=O
Applications
6-Methoxy-2-naphthaldehyde is used as a diagnostic reagent in tumor studies involving aldehyde dehydrogenase enzymes. In addition it is used in organic synthesis reactions forming fluorescent substrates for inhibition studies relating to hypertension and vascular inflammation. It is also used as intermediate of Nabumetone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to air. Incompatible with oxidizing agents.
Melting Point 80°C to 82°C
Quantity 100g
Beilstein 1945989
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 6-Methoxy-2-naphthaldehyde