Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methoxy-2-naphthonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
67886-70-8
C12H9NO
183.21
SWEFYGCKCMJSPD-UHFFFAOYSA-N
6-methoxy-2-naphthonitrile,2-cyano-6-methoxynaphthalene,6-methoxy-2-cyanonaphthalene,6-methoxy-2-naphthalenecarbonitrile,6-methoxy-naphthalene-2-carbonitrile,2-naphthalenecarbonitrile,6-methoxy,acmc-20anxg,2-cyan-6-methoxy-naphthalin,2-naphthonitrile, 6-methoxy
349173
6-methoxynaphthalene-2-carbonitrile
COC1=CC2=C(C=C1)C=C(C=C2)C#N
67886-70-8
C12H9NO
183.21
SWEFYGCKCMJSPD-UHFFFAOYSA-N
6-methoxy-2-naphthonitrile,2-cyano-6-methoxynaphthalene,6-methoxy-2-cyanonaphthalene,6-methoxy-2-naphthalenecarbonitrile,6-methoxy-naphthalene-2-carbonitrile,2-naphthalenecarbonitrile,6-methoxy,acmc-20anxg,2-cyan-6-methoxy-naphthalin,2-naphthonitrile, 6-methoxy
349173
6-methoxynaphthalene-2-carbonitrile
COC1=CC2=C(C=C1)C=C(C=C2)C#N
Packaging Glass Bottle
Boiling Point 205.0°C to 208.0°C (14.0mmHg)
Quantity 1g
Infrared Spectrum Authentic
Color White to Yellow
Melting Point 106.0°C to 107.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Linear Formula CH3OC10H6CN
Beilstein 10,III,1099
Formula Weight 183.21
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 6-Methoxy-2-naphthonitrile
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning