Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Methoxy-2-naphthylacetic acid, 99%

Click to view available options
Quantity:
5mg
10mg
50mg
23981-47-7
C13H12O3
216.236
MFCD00033105
PHJFLPMVEFKEPL-UHFFFAOYSA-N
2-6-methoxynaphthalen-2-yl acetic acid,6-methoxy-2-naphthylacetic acid,6-mnaa,6-methoxy-2-naphthaleneacetic acid,6-methoxy-2-naphthyl acetic acid,unii-9h7k3yod7q,6-methoxynaphthalene-2-acetic acid,2-6-methoxy-2-naphthyl acetic acid,6-metossi-2-naphtil acetico italian,9h7k3yod7q
32176
CHEBI:35628
2-(6-methoxynaphthalen-2-yl)acetic acid
COC1=CC2=C(C=C1)C=C(C=C2)CC(=O)O
23981-47-7
C13H12O3
216.236
MFCD00033105
PHJFLPMVEFKEPL-UHFFFAOYSA-N
2-6-methoxynaphthalen-2-yl acetic acid,6-methoxy-2-naphthylacetic acid,6-mnaa,6-methoxy-2-naphthaleneacetic acid,6-methoxy-2-naphthyl acetic acid,unii-9h7k3yod7q,6-methoxynaphthalene-2-acetic acid,2-6-methoxy-2-naphthyl acetic acid,6-metossi-2-naphtil acetico italian,9h7k3yod7q
32176
CHEBI:35628
2-(6-methoxynaphthalen-2-yl)acetic acid
COC1=CC2=C(C=C1)C=C(C=C2)CC(=O)O
Quantity 50mg
Melting Point 171°C to 173°C
Formula Weight 216.24
Physical Form Crystalline Solid
Percent Purity 99%
Chemical Name or Material 6-Methoxy-2-naphthylacetic acid

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : QJ1045000