Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methoxy-2-naphthylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Acros Organics

Click to view available options
Quantity:
50mL
Packaging:
AcroSeal™ Glass Bottle
38046-82-1,109-99-9
C11H9BrMgO
261.4
MFCD01311488
DMMNAMUAQVNNBC-UHFFFAOYSA-M
6-methoxy-2-naphthylmagnesium bromide,sogqgmfueumyjd-uhfffaoysa-m,6-methoxynaphthalen-2-ylmagnesium bromide,4-fluorobenzylmagnesium chloride 0.25m&,6-methoxy-2-naphthylmagnesium bromide, 0.5m in thf,6-methoxy-2-naphthylmagnesium bromide 0.5 m in tetrahydrofuran,6-methoxy-2-naphthylmagnesium bromide solution, 0.5 m in thf
11242453
magnesium;6-methoxy-2H-naphthalen-2-ide;bromide
COC1=CC2=CC=[C-]C=C2C=C1.[Mg+2].[Br-]
38046-82-1,109-99-9
C11H9BrMgO
261.4
MFCD01311488
DMMNAMUAQVNNBC-UHFFFAOYSA-M
6-methoxy-2-naphthylmagnesium bromide,sogqgmfueumyjd-uhfffaoysa-m,6-methoxynaphthalen-2-ylmagnesium bromide,4-fluorobenzylmagnesium chloride 0.25m&,6-methoxy-2-naphthylmagnesium bromide, 0.5m in thf,6-methoxy-2-naphthylmagnesium bromide 0.5 m in tetrahydrofuran,6-methoxy-2-naphthylmagnesium bromide solution, 0.5 m in thf
11242453
magnesium;6-methoxy-2H-naphthalen-2-ide;bromide
COC1=CC2=CC=[C-]C=C2C=C1.[Mg+2].[Br-]
Specific Gravity 0.966
Packaging AcroSeal™ Glass Bottle
Quantity 50mL
Color Brown
Density 0.9660g/mL
Flash Point −25°C
CAS Min % 10.82,83.76
CAS Max % 16.24,89.18
Solubility Information Solubility in water: reacts
Formula Weight 261.4
Physical Form Liquid
Chemical Name or Material 6-Methoxy-2-naphthylmagnesium bromide
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger