Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methoxy-3-pyridinecarboxaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
65873-72-5
C7H7NO2
137.14
MFCD02683446
CTAIEPPAOULMFY-UHFFFAOYSA-N
6-methoxynicotinaldehyde,6-methoxy-3-pyridinecarboxaldehyde,5-formyl-2-methoxypyridine,2-methoxypyridine-5-carboxaldehyde,5-formyl-2-methoxy-pyridine,6-methoxy-pyridine-3-carbaldehyde,6-methoxypyridine-3-carboxaldehyde,3-pyridinecarboxaldehyde, 6-methoxy,6-methoxy-3-nicotinaldehyde,pubchem5152
3364576
6-methoxypyridine-3-carbaldehyde
COC1=NC=C(C=C1)C=O
65873-72-5
C7H7NO2
137.14
MFCD02683446
CTAIEPPAOULMFY-UHFFFAOYSA-N
6-methoxynicotinaldehyde,6-methoxy-3-pyridinecarboxaldehyde,5-formyl-2-methoxypyridine,2-methoxypyridine-5-carboxaldehyde,5-formyl-2-methoxy-pyridine,6-methoxy-pyridine-3-carbaldehyde,6-methoxypyridine-3-carboxaldehyde,3-pyridinecarboxaldehyde, 6-methoxy,6-methoxy-3-nicotinaldehyde,pubchem5152
3364576
6-methoxypyridine-3-carbaldehyde
COC1=NC=C(C=C1)C=O
Packaging Glass bottle
Flash Point >110°C
Infrared Spectrum Authentic
Melting Point 46°C to 54°C
Color White to Yellow
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Solubility Information Solubility in water: slightly soluble
Formula Weight 137.14
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 6-Methoxy-3-pyridinecarboxaldehyde
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medi

GHS Signal Word: Warning