Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Methoxyflavanone, 98%

Click to view available options
Quantity:
1g
5g
25g
3034-04-6
C16H14O3
254.29
MFCD00017484
YURQMHCZHLMHIB-UHFFFAOYNA-N
6-methoxyflavanone,6-methoxy-2-phenylchroman-4-one,6-methoxy-2-phenyl-chroman-4-one,6-methoxy-2-phenyl-2,3-dihydro-1-benzopyran-4-one,flavanone, 6-methoxy,acmc-1adv8,6-methoxy-2-phenyl-2,3-dihydro-4h-chromen-4-one,6-methoxy-2-phenyl-2,3-dihydro-4h-chromen-4-one #,2,3-dihydro-6-methoxy-2-phenyl-4h-1-benzopyran-4-one,4h-1-benzopyran-4-one, 2,3-dihydro-6-methoxy-2-phenyl
97860
6-methoxy-2-phenyl-2,3-dihydrochromen-4-one
COC1=CC2=C(C=C1)OC(CC2=O)C3=CC=CC=C3
3034-04-6
C16H14O3
254.29
MFCD00017484
YURQMHCZHLMHIB-UHFFFAOYNA-N
6-methoxyflavanone,6-methoxy-2-phenylchroman-4-one,6-methoxy-2-phenyl-chroman-4-one,6-methoxy-2-phenyl-2,3-dihydro-1-benzopyran-4-one,flavanone, 6-methoxy,acmc-1adv8,6-methoxy-2-phenyl-2,3-dihydro-4h-chromen-4-one,6-methoxy-2-phenyl-2,3-dihydro-4h-chromen-4-one #,2,3-dihydro-6-methoxy-2-phenyl-4h-1-benzopyran-4-one,4h-1-benzopyran-4-one, 2,3-dihydro-6-methoxy-2-phenyl
97860
6-methoxy-2-phenyl-2,3-dihydrochromen-4-one
COC1=CC2=C(C=C1)OC(CC2=O)C3=CC=CC=C3
Quantity 25g
Melting Point 141°C to 143°C
Beilstein 248912
Formula Weight 254.29
Percent Purity 98%
Chemical Name or Material 6-Methoxyflavanone