Alfa Aesar™ 6-Methoxyflavanone, 98%

Manufacturer: Alfa Aesar™ B2524003

Quantity 1g

 View more versions of this product

Catalog No. AAB2524003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3034-04-6
    Molecular Formula C16H14O3
    Formula Weight 254.29
    MDL Number MFCD00017484
    Beilstein 248912
    Chemical Name or Material 6-Methoxyflavanone

    Specifications

    Formula Weight 254.29
    Melting Point 141° to 143°C
    Quantity 1g
    Molecular Formula C16H14O3
    MDL Number MFCD00017484
    CAS 3034-04-6
    Beilstein 248912
    Chemical Name or Material 6-Methoxyflavanone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit