Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Methoxyindole-2-carboxylic acid, 95%

Click to view available options
Quantity:
1g
5g
16732-73-3
C10H9NO3
191.186
MFCD01548823
XNBGANWAZJWOHS-UHFFFAOYSA-N
6-methoxyindole-2-carboxylic acid,1h-indole-2-carboxylic acid, 6-methoxy,pubchem1703,zlchem 1017,6-methoxindole-2-carboxylic acid,indole-2-carboxylic acid, 6-methoxy,asinex-reag bas 06489245,6-methoxy-1 h-indole-2-carboxylic acid,6-methoxyindole-2-carboxylic acid,#
410327
6-methoxy-1H-indole-2-carboxylic acid
COC1=CC2=C(C=C1)C=C(N2)C(=O)O
16732-73-3
C10H9NO3
191.186
MFCD01548823
XNBGANWAZJWOHS-UHFFFAOYSA-N
6-methoxyindole-2-carboxylic acid,1h-indole-2-carboxylic acid, 6-methoxy,pubchem1703,zlchem 1017,6-methoxindole-2-carboxylic acid,indole-2-carboxylic acid, 6-methoxy,asinex-reag bas 06489245,6-methoxy-1 h-indole-2-carboxylic acid,6-methoxyindole-2-carboxylic acid,#
410327
6-methoxy-1H-indole-2-carboxylic acid
COC1=CC2=C(C=C1)C=C(N2)C(=O)O
Melting Point 198°C to 203°C
Quantity 5g
Beilstein 156983
Formula Weight 191.19
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 6-Methoxyindole-2-carboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning