Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methoxyindole, 98%, ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass Bottle
3189-13-7
C9H9NO
147.177
MFCD00022780
QJRWYBIKLXNYLF-UHFFFAOYSA-N
6-methoxyindole,1h-indole, 6-methoxy,indole, 6-methoxy,1h-indol-6-yl methyl ether,6-methoxy indole,pubchem1701,acmc-209hph,opera_id_1903,6-methyloxy-1h-indole
76659
6-methoxy-1H-indole
COC1=CC2=C(C=C1)C=CN2
3189-13-7
C9H9NO
147.177
MFCD00022780
QJRWYBIKLXNYLF-UHFFFAOYSA-N
6-methoxyindole,1h-indole, 6-methoxy,indole, 6-methoxy,1h-indol-6-yl methyl ether,6-methoxy indole,pubchem1701,acmc-209hph,opera_id_1903,6-methyloxy-1h-indole
76659
6-methoxy-1H-indole
COC1=CC2=C(C=C1)C=CN2
Packaging Glass Bottle
Infrared Spectrum Authentic
Color White
Quantity 500mg
Melting Point 90.0°C to 94.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 21, 22
Formula Weight 147.18
Physical Form Shiny Crystals
Percent Purity 98%
Chemical Name or Material 6-Methoxyindole, 98%
Show More Show Less