Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methoxyquinaldine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1078-28-0
C11H11NO
173.21
MFCD00006761
NAGJQQFMJKMXJQ-UHFFFAOYSA-N
6-methoxyquinaldine,2-methyl-6-methoxyquinoline,quinoline, 6-methoxy-2-methyl,quinaldine, 6-methoxy,6-methoxy-2-methyl-quinoline,quinoline,6-methoxy-2-methyl,pubchem5910,acmc-2098xp,2-methy-6-methoxyquinolinel
70648
6-methoxy-2-methylquinoline
CC1=NC2=C(C=C1)C=C(C=C2)OC
1078-28-0
C11H11NO
173.21
MFCD00006761
NAGJQQFMJKMXJQ-UHFFFAOYSA-N
6-methoxyquinaldine,2-methyl-6-methoxyquinoline,quinoline, 6-methoxy-2-methyl,quinaldine, 6-methoxy,6-methoxy-2-methyl-quinoline,quinoline,6-methoxy-2-methyl,pubchem5910,acmc-2098xp,2-methy-6-methoxyquinolinel
70648
6-methoxy-2-methylquinoline
CC1=NC2=C(C=C1)C=C(C=C2)OC
Color Beige to Gray
Melting Point 61°C to 65°C
Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 173.21
Physical Form Fluffy Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 6-Methoxyquinaldine
Show More Show Less