Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Methoxytryptamine, 95%

Click to view available options
Quantity:
1g
5g
3610-36-4
C11H14N2O
190.246
MFCD00005663
VOCGEKMEZOPDFP-UHFFFAOYSA-N
6-methoxytryptamine,2-6-methoxy-1h-indol-3-yl ethanamine,3-2-aminoethyl-6-methoxyindole,2-6-methoxy-1h-indol-3-yl-ethylamine,1h-indole-3-ethanamine, 6-methoxy,2-2-aminoethyl-5-methoxyindole,1h-indole-3-ethanamine,6-methoxy,2-6-methoxy-1h-indol-3-yl ethan-1-amine,2-6-methoxyindol-3-yl ethylamine,6-methoxy tryptamine
17654
2-(6-methoxy-1H-indol-3-yl)ethanamine
COC1=CC2=C(C=C1)C(=CN2)CCN
3610-36-4
C11H14N2O
190.246
MFCD00005663
VOCGEKMEZOPDFP-UHFFFAOYSA-N
6-methoxytryptamine,2-6-methoxy-1h-indol-3-yl ethanamine,3-2-aminoethyl-6-methoxyindole,2-6-methoxy-1h-indol-3-yl-ethylamine,1h-indole-3-ethanamine, 6-methoxy,2-2-aminoethyl-5-methoxyindole,1h-indole-3-ethanamine,6-methoxy,2-6-methoxy-1h-indol-3-yl ethan-1-amine,2-6-methoxyindol-3-yl ethylamine,6-methoxy tryptamine
17654
2-(6-methoxy-1H-indol-3-yl)ethanamine
COC1=CC2=C(C=C1)C(=CN2)CCN
Melting Point 146°C to 147°C
Odor Odorless
Quantity 5g
Beilstein 7948
Formula Weight 190.25
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 6-Methoxytryptamine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C