Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methoxytryptamine, 99%, ACROS Organics™

Click to view available options
Quantity:
50mg
250mg
1g
5g
Packaging:
Glass bottle
3610-36-4
C11H14N2O
190.24
MFCD00005663
VOCGEKMEZOPDFP-UHFFFAOYSA-N
6-methoxytryptamine,2-6-methoxy-1h-indol-3-yl ethanamine,3-2-aminoethyl-6-methoxyindole,2-6-methoxy-1h-indol-3-yl-ethylamine,1h-indole-3-ethanamine, 6-methoxy,2-2-aminoethyl-5-methoxyindole,1h-indole-3-ethanamine,6-methoxy,2-6-methoxy-1h-indol-3-yl ethan-1-amine,2-6-methoxyindol-3-yl ethylamine,6-methoxy tryptamine
17654
2-(6-methoxy-1H-indol-3-yl)ethanamine
COC1=CC2=C(C=C1)C(=CN2)CCN
3610-36-4
C11H14N2O
190.24
MFCD00005663
VOCGEKMEZOPDFP-UHFFFAOYSA-N
6-methoxytryptamine,2-6-methoxy-1h-indol-3-yl ethanamine,3-2-aminoethyl-6-methoxyindole,2-6-methoxy-1h-indol-3-yl-ethylamine,1h-indole-3-ethanamine, 6-methoxy,2-2-aminoethyl-5-methoxyindole,1h-indole-3-ethanamine,6-methoxy,2-6-methoxy-1h-indol-3-yl ethan-1-amine,2-6-methoxyindol-3-yl ethylamine,6-methoxy tryptamine
17654
2-(6-methoxy-1H-indol-3-yl)ethanamine
COC1=CC2=C(C=C1)C(=CN2)CCN
Packaging Glass bottle
Quantity 1g
Melting Point 143.0°C to 146.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Solubility Information Solubility in water: moderately soluble. Other solubilities: soluble in ethanol
Formula Weight 190.24
Percent Purity 99%
Chemical Name or Material 6-Methoxytryptamine, 99%

GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 222-778-2