Alfa Aesar™ 6-Methyl-1-heptylamine, 97%

Manufacturer: Alfa Aesar™ B2474614

Quantity 25g

 View more versions of this product

Catalog No. AAB2474614

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1321-35-3
    Molecular Formula (CH3)2CH(CH2)5NH2
    Formula Weight 129.25
    MDL Number MFCD00272272
    UN Number UN2735
    Synonym 1-Amino-6-methylheptane; Isooctylamine
    Chemical Name or Material 6-Methyl-1-heptylamine

    Specifications

    Formula Weight 129.25
    Health Hazard 1 Danger
    Quantity 25g
    Molecular Formula (CH3)2CH(CH2)5NH2
    MDL Number MFCD00272272
    CAS 1321-35-3
    UN Number UN2735
    Synonym 1-Amino-6-methylheptane; Isooctylamine
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Chemical Name or Material 6-Methyl-1-heptylamine
    Assay 97%

    Description

    Applications
    Used as pharmaceutical intermediate.

    Notes
    Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit