Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methyl-2-pyridinecarboxaldehyde 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
1122-72-1
C7H7NO
121.14
MFCD00006291
AHISYUZBWDSPQL-UHFFFAOYSA-N
6-methyl-2-pyridinecarboxaldehyde,6-methylpicolinaldehyde,2-formyl-6-methylpyridine,6-methylpyridine-2-carboxaldehyde,2-pyridinecarboxaldehyde, 6-methyl,6-methylpyridine-2-aldehyde,2-formyl-6-picoline,6-methyl-2-pyridinecarbaldehyde,6-methyl-2-picolinaldehyde,6-methylpyridyl-2-aldehyde
70737
6-methylpyridine-2-carbaldehyde
CC1=CC=CC(=N1)C=O
1122-72-1
C7H7NO
121.14
MFCD00006291
AHISYUZBWDSPQL-UHFFFAOYSA-N
6-methyl-2-pyridinecarboxaldehyde,6-methylpicolinaldehyde,2-formyl-6-methylpyridine,6-methylpyridine-2-carboxaldehyde,2-pyridinecarboxaldehyde, 6-methyl,6-methylpyridine-2-aldehyde,2-formyl-6-picoline,6-methyl-2-pyridinecarbaldehyde,6-methyl-2-picolinaldehyde,6-methylpyridyl-2-aldehyde
70737
6-methylpyridine-2-carbaldehyde
CC1=CC=CC(=N1)C=O
Melting Point 31°C to 33°C
Flash Point 68°C
Refractive Index 1.527
Color Brown to Yellow
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH(C5H3N)COH
Solubility Information Solubility in water: 750g/L water (20°C). Other solubilities: soluble in ether and acetone
Formula Weight 121.14
Physical Form Low Melting Solid
Percent Purity ≥96.0%
Chemical Name or Material 6-Methyl-2-pyridinecarboxaldehyde, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LIGHT SENSITIVE

EINECSNumber : 214-359-8

TSCA : TSCA