Alfa Aesar™ 6-Methyl-2-pyridinemethanol, 98%

Manufacturer: Alfa Aesar™ H2679403

Quantity 1g

 View more versions of this product

Catalog No. AAH2679403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1122-71-0
    Molecular Formula C7H9NO
    Formula Weight 123.15
    MDL Number MFCD00023520
    Synonym alpha-Hydroxy-2,6-lutidine; 6-Hydroxymethyl-2-picoline
    Chemical Name or Material 6-Methyl-2-pyridinemethanol

    Specifications

    Formula Weight 123.15
    Flash Point 109°C (228°F)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C7H9NO
    MDL Number MFCD00023520
    CAS 1122-71-0
    Synonym alpha-Hydroxy-2,6-lutidine; 6-Hydroxymethyl-2-picoline
    Health Hazard 1 Warning
    Melting Point 32° to 34°C
    Boiling Point 105° to 108°C (12mmHg)
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 6-Methyl-2-pyridinemethanol
    Assay 98%

    Description

    Applications
    Use as primary and secondary intermediate.

    Solubility
    Low soluble in water, high soluble in ethanol, acetic acid. Soluble in benzene, toluene.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, strong reducing agents. Store under an inert atmosphere.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit