Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methyl-2-pyridinemethanol, 98%, Thermo Scientific Chemicals

Catalog Number p-7029341
Click to view available options
Quantity:
5 g
25 g
1122-71-0
C7H9NO
123.16
MFCD00023520
JLVBSBMJQUMAMW-UHFFFAOYSA-N
6-methyl-2-pyridinemethanol, 6-methylpyridin-2-yl methanol, 2-pyridinemethanol, 6-methyl, 6-methyl-2-pyridylmethanol, 6-methylpyridine-2-methanol, 2-hydroxymethyl-6-methylpyridine, 6-methyl-2-pyridyl methan-1-ol, pubchem20959, acmc-2099dw, 6-methyl-2-pyridylcarbinol
70736
(6-methylpyridin-2-yl)methanol
CC1=CC=CC(CO)=N1
1122-71-0
C7H9NO
123.16
MFCD00023520
JLVBSBMJQUMAMW-UHFFFAOYSA-N
6-methyl-2-pyridinemethanol, 6-methylpyridin-2-yl methanol, 2-pyridinemethanol, 6-methyl, 6-methyl-2-pyridylmethanol, 6-methylpyridine-2-methanol, 2-hydroxymethyl-6-methylpyridine, 6-methyl-2-pyridyl methan-1-ol, pubchem20959, acmc-2099dw, 6-methyl-2-pyridylcarbinol
70736
(6-methylpyridin-2-yl)methanol
CC1=CC=CC(CO)=N1

Use as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Use as primary and secondary intermediate.

Solubility
Low soluble in water, high soluble in ethanol, acetic acid. Soluble in benzene, toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, strong reducing agents. Store under an inert atmosphere.
TRUSTED_SUSTAINABILITY
Melting Point 32°C to 34°C
Boiling Point 105°C to 108°C (12 mmHg)
Flash Point 109°C (228°F)
Quantity 5 g
Solubility Information Low soluble in water,high soluble in ethanol,acetic acid. Soluble in benzene,toluene.
Formula Weight 123.15
Percent Purity 98%
Chemical Name or Material 6-Methyl-2-pyridinemethanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-358-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit