Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Methylcoumarin, 99%

Click to view available options
Quantity:
50g
100g
250g
92-48-8
C10H8O2
160.172
MFCD00006875
FXFYOPQLGGEACP-UHFFFAOYSA-N
6-methylcoumarin,6-methylbenzopyrone,6-methyl-2h-chromen-2-one,toncarine,6-methyl coumarin,methyl coumarin,6-methylcoumarinic anhydride,coumarin, 6-methyl,6-methyl-1,2-benzopyrone,6-methylcumarin
7092
CHEBI:563586
6-methylchromen-2-one
CC1=CC2=C(C=C1)OC(=O)C=C2
92-48-8
C10H8O2
160.172
MFCD00006875
FXFYOPQLGGEACP-UHFFFAOYSA-N
6-methylcoumarin,6-methylbenzopyrone,6-methyl-2h-chromen-2-one,toncarine,6-methyl coumarin,methyl coumarin,6-methylcoumarinic anhydride,coumarin, 6-methyl,6-methyl-1,2-benzopyrone,6-methylcumarin
7092
CHEBI:563586
6-methylchromen-2-one
CC1=CC2=C(C=C1)OC(=O)C=C2
Boiling Point 303°C (725mmHg)
Melting Point 74°C to 77°C
Quantity 250g
Beilstein 4222
Formula Weight 160.17
Percent Purity 99%
Chemical Name or Material 6-Methylcoumarin

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c

H301+H311+H331

EINECSNumber : 202-158-8

RTECSNumber : GN7792000

TSCA : Yes

Recommended Storage : Ambient temperatures