Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methylindole, 99%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass Bottle
3420-02-8
C9H9N
131.18
MFCD00005682
ONYNOPPOVKYGRS-UHFFFAOYSA-N
n-boc-aniline,tert-butyl phenylcarbamate,n-boc aniline,t-butyl carbanilate,tert-butyl carbanilate,carbanilic acid, tert-butyl ester,carbamic acid, phenyl-, 1,1-dimethylethyl ester,n-tert-butoxycarbonyl aniline,tert-butyl n-phenylurethane,carbanilic acid tert-butyl ester
137930
tert-butyl N-phenylcarbamate
CC1=CC=C2C=CNC2=C1
3420-02-8
C9H9N
131.18
MFCD00005682
ONYNOPPOVKYGRS-UHFFFAOYSA-N
n-boc-aniline,tert-butyl phenylcarbamate,n-boc aniline,t-butyl carbanilate,tert-butyl carbanilate,carbanilic acid, tert-butyl ester,carbamic acid, phenyl-, 1,1-dimethylethyl ester,n-tert-butoxycarbonyl aniline,tert-butyl n-phenylurethane,carbanilic acid tert-butyl ester
137930
tert-butyl N-phenylcarbamate
CC1=CC=C2C=CNC2=C1
Boiling Point 112°C (5.0mmHg)
Flash Point >112°C
Packaging Glass Bottle
Specific Gravity 1.05
Refractive Index 1.6060 to 1.608
Density 1.0500g/mL
Quantity 250mg
Color Yellow to Brown
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 20, IV, 3214
Formula Weight 131.18
Physical Form Liquid After Melting
Percent Purity ≥98.5%
Chemical Name or Material 6-Methylindole
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : This product darkens on exposure to light