Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Methylpyridine-3-carboxaldehyde, 96%

Click to view available options
Quantity:
250mg
1g
5g
53014-84-9
C7H7NO
121.139
MFCD08272279
IMWMEIWYPWVABQ-UHFFFAOYSA-N
6-methylnicotinaldehyde,2-methyl-5-formylpyridine,6-methyl-pyridine-3-carbaldehyde,3-formyl-6-methyl-pyridine,5-formyl-2-methylpyridine,5-formyl-2-picoline,6-methylpyridine-3-carboxaldehyde,3-pyridinecarboxaldehyde, 6-methyl,3-formyl-6-methylpyridine,6-methylpyridyl-3-carboxaldehyde
10192566
6-methylpyridine-3-carbaldehyde
CC1=NC=C(C=C1)C=O
53014-84-9
C7H7NO
121.139
MFCD08272279
IMWMEIWYPWVABQ-UHFFFAOYSA-N
6-methylnicotinaldehyde,2-methyl-5-formylpyridine,6-methyl-pyridine-3-carbaldehyde,3-formyl-6-methyl-pyridine,5-formyl-2-methylpyridine,5-formyl-2-picoline,6-methylpyridine-3-carboxaldehyde,3-pyridinecarboxaldehyde, 6-methyl,3-formyl-6-methylpyridine,6-methylpyridyl-3-carboxaldehyde
10192566
6-methylpyridine-3-carbaldehyde
CC1=NC=C(C=C1)C=O
Sensitivity Moisture Sensitive
Boiling Point 95°C (20mmHg)
Refractive Index 1.5460
Density 1.084g/mL
Quantity 5g
Beilstein 107496
Formula Weight 121.14
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 6-Methylpyridine-3-carboxaldehyde