Alfa Aesar™ 6-Methylquinoline, 98%

Manufacturer: Alfa Aesar™ B2189206

Quantity 5g

 View more versions of this product

Catalog No. AAB2189206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 91-62-3
    Molecular Formula C10H9N
    Formula Weight 143.19
    MDL Number MFCD00006804
    Beilstein 110336
    Synonym p-Toluquinoline
    Chemical Name or Material 6-Methylquinoline

    Specifications

    Formula Weight 143.19
    MDL Number MFCD00006804
    Melting Point −22°C
    Health Hazard 3 GHS P Statement
    P280-P281-P301+P310-P305+P351+P338-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Molecular Formula C10H9N
    CAS 91-62-3
    Beilstein 110336
    Synonym p-Toluquinoline
    Flash Point >110°C (230°F)
    Health Hazard 1 Danger
    RTECS Number VC0550000
    Quantity 5g
    Boiling Point 257° to 259°C
    Health Hazard 2 GHS H Statement
    H301-H341-H315-H319
    Toxic if swallowed.
    Suspected of causing genetic defects.
    Causes skin irritation.
    Causes serious eye irritation.
    Chemical Name or Material 6-Methylquinoline
    Assay 98%

    Description

    Applications
    6-Methylquinoline can be used as primary carbon source in culture of Pseudomonas putida QP1. 6-Methylquinoline was used in the synthesis of fluorescent halide-sensitive quinolinium dyes and fluorescent probes for determination of chloride in biological systems.

    Solubility
    Not miscible or difficult to mix with water.

    Notes
    Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    GHS H Statement
    H301-H341-H315-H319
    Toxic if swallowed.
    Suspected of causing genetic defects.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P281-P301+P310-P305+P351+P338-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit