Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Methyluracil, 97%

Click to view available options
Quantity:
100g
500g
2500g
626-48-2
C5H6N2O2
126.115
MFCD00006028
SHVCSCWHWMSGTE-UHFFFAOYSA-N
6-methyluracil,2,4-dihydroxy-6-methylpyrimidine,pseudothymine,6-methylpyrimidine-2,4 1h,3h-dione,4-methyluracil,6-methylpyrimidine-2,4-diol,2,4 1h,3h-pyrimidinedione, 6-methyl,uracil, 6-methyl,6-methyl-2,4 1h,3h-pyrimidinedione,2,4-pyrimidinediol, 6-methyl
12283
CHEBI:74733
6-methyl-1H-pyrimidine-2,4-dione
CC1=CC(=O)NC(=O)N1
626-48-2
C5H6N2O2
126.115
MFCD00006028
SHVCSCWHWMSGTE-UHFFFAOYSA-N
6-methyluracil,2,4-dihydroxy-6-methylpyrimidine,pseudothymine,6-methylpyrimidine-2,4 1h,3h-dione,4-methyluracil,6-methylpyrimidine-2,4-diol,2,4 1h,3h-pyrimidinedione, 6-methyl,uracil, 6-methyl,6-methyl-2,4 1h,3h-pyrimidinedione,2,4-pyrimidinediol, 6-methyl
12283
CHEBI:74733
6-methyl-1H-pyrimidine-2,4-dione
CC1=CC(=O)NC(=O)N1
Melting Point 317°C to 320°C
Quantity 2500g
Beilstein 115647
Merck Index 14,6133
Formula Weight 126.12
Percent Purity 97%
Chemical Name or Material 6-Methyluracil

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : YR0700000

TSCA : TSCA