Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(6-morpholinopyrid-2-yl)methylamine, 95%, Maybridge™

Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
868755-52-6
C10H15N3O
193.25
MFCD08435862
JRKWUCQBVPZBBP-UHFFFAOYSA-N
6-morpholinopyrid-2-yl methylamine,6-morpholin-4-yl pyridin-2-yl methanamine,2-pyridinemethanamine,6-4-morpholinyl,6-morpholin-4-ylpyridin-2-yl methylamine,1-6-morpholin-4-yl pyridin-2-yl methanamine,6-morpholinopyridin-2-yl methanamine,6-morpholinopyridin-2-yl methylamine,6-morpholin-4-yl-2-pyridyl methylamine,2-aminomethyl-6-morpholin-4-yl pyridine
18525737
(6-morpholin-4-ylpyridin-2-yl)methanamine
C1COCCN1C2=CC=CC(=N2)CN
868755-52-6
C10H15N3O
193.25
MFCD08435862
JRKWUCQBVPZBBP-UHFFFAOYSA-N
6-morpholinopyrid-2-yl methylamine,6-morpholin-4-yl pyridin-2-yl methanamine,2-pyridinemethanamine,6-4-morpholinyl,6-morpholin-4-ylpyridin-2-yl methylamine,1-6-morpholin-4-yl pyridin-2-yl methanamine,6-morpholinopyridin-2-yl methanamine,6-morpholinopyridin-2-yl methylamine,6-morpholin-4-yl-2-pyridyl methylamine,2-aminomethyl-6-morpholin-4-yl pyridine
18525737
(6-morpholin-4-ylpyridin-2-yl)methanamine
C1COCCN1C2=CC=CC(=N2)CN
Packaging Amber Glass Bottle
Quantity 250mg
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 193.25
Percent Purity 95%
Chemical Name or Material (6-Morpholinopyrid-2-yl)methylamine

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger