Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Nitro-2H-1,4-benzothiazin-3(4H)-one, 97%, Thermo Scientific™

Click to view available options
Quantity:
250 mg
1 g
21762-74-3
C8H6N2O3S
210.21
MFCD00456556
BALSKAVZBPPKCS-UHFFFAOYSA-N
6-nitro-2h-benzo b 1,4 thiazin-3 4h-one, 6-nitro-2h-1,4-benzothiazin-3 4h-one, 2h-1,4-benzothiazin-3 4h-one, 6-nitro, 6-nitro-2,4-dihydro-1,4-benzothiazin-3-one, 2h-1,4-benzothiazin-3 4h-one,6-nitro, 6-nitro-2h-benzo b-1,4 thiazin-3 4h-one, 6-nitro-3,4-dihydro-2h-1,4-benzothiazin-3-one
4182356
6-nitro-3,4-dihydro-2H-1,4-benzothiazin-3-one
[O-][N+](=O)C1=CC2=C(SCC(=O)N2)C=C1
21762-74-3
C8H6N2O3S
210.21
MFCD00456556
BALSKAVZBPPKCS-UHFFFAOYSA-N
6-nitro-2h-benzo b 1,4 thiazin-3 4h-one, 6-nitro-2h-1,4-benzothiazin-3 4h-one, 2h-1,4-benzothiazin-3 4h-one, 6-nitro, 6-nitro-2,4-dihydro-1,4-benzothiazin-3-one, 2h-1,4-benzothiazin-3 4h-one,6-nitro, 6-nitro-2h-benzo b-1,4 thiazin-3 4h-one, 6-nitro-3,4-dihydro-2h-1,4-benzothiazin-3-one
4182356
6-nitro-3,4-dihydro-2H-1,4-benzothiazin-3-one
[O-][N+](=O)C1=CC2=C(SCC(=O)N2)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Quantity 250 mg
Formula Weight 210.21
Percent Purity 97%
Chemical Name or Material 6-Nitro-2H-1,4-benzothiazin-3(4H)-one

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit