Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Nitrophthalide, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
610-93-5
C8H5NO4
179.131
MFCD00033529
RNWGZXAHUPFXLL-UHFFFAOYSA-N
6-nitrophthalide,6-nitroisobenzofuran-1 3h-one,6-nitro-3h-isobenzofuran-1-one,6-nitro-1,3-dihydroisobenzofuran-1-one,6-nitro-1 3h-isobenzofuranone,6-nitro-2-benzofuran-1 3h-one,1 3h-isobenzofuranone, 6-nitro,6-nitro-3-hydroisobenzofuran-1-one,6-nitro-1,3-dihydro-2-benzofuran-1-one,6-nitrophtalide
223584
6-nitro-3H-2-benzofuran-1-one
C1C2=C(C=C(C=C2)[N+](=O)[O-])C(=O)O1
610-93-5
C8H5NO4
179.131
MFCD00033529
RNWGZXAHUPFXLL-UHFFFAOYSA-N
6-nitrophthalide,6-nitroisobenzofuran-1 3h-one,6-nitro-3h-isobenzofuran-1-one,6-nitro-1,3-dihydroisobenzofuran-1-one,6-nitro-1 3h-isobenzofuranone,6-nitro-2-benzofuran-1 3h-one,1 3h-isobenzofuranone, 6-nitro,6-nitro-3-hydroisobenzofuran-1-one,6-nitro-1,3-dihydro-2-benzofuran-1-one,6-nitrophtalide
223584
6-nitro-3H-2-benzofuran-1-one
C1C2=C(C=C(C=C2)[N+](=O)[O-])C(=O)O1
Melting Point 140°C to 145°C
Quantity 1g
Color Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 17, 313
Formula Weight 179.13
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 6-Nitrophthalide, 97%
Show More Show Less