Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Nitropiperonyl alcohol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
15341-08-9
C8H7NO5
197.146
MFCD00005825
XSKQKDTZQNFCCB-UHFFFAOYSA-N
6-nitropiperonyl alcohol,6-nitrobenzo d 1,3 dioxol-5-yl methanol,3,4-methylenedioxy-6-nitrobenzyl alcohol,6-nitro-3,4-methylenedioxybenzyl alcohol,6-nitro-1,3-benzodioxol-5-yl methanol,6-nitro-2h-1,3-benzodioxol-5-yl methanol,6-nitro-1,3-benzodioxole-5-methanol,1,3-benzodioxole-5-methanol,6-nitro,1,3-benzodioxole-5-methanol, 6-nitro,6-nitro-1,3-benzodioxol-5-yl methanol #
519111
(6-nitro-1,3-benzodioxol-5-yl)methanol
C1OC2=C(O1)C=C(C(=C2)CO)[N+](=O)[O-]
15341-08-9
C8H7NO5
197.146
MFCD00005825
XSKQKDTZQNFCCB-UHFFFAOYSA-N
6-nitropiperonyl alcohol,6-nitrobenzo d 1,3 dioxol-5-yl methanol,3,4-methylenedioxy-6-nitrobenzyl alcohol,6-nitro-3,4-methylenedioxybenzyl alcohol,6-nitro-1,3-benzodioxol-5-yl methanol,6-nitro-2h-1,3-benzodioxol-5-yl methanol,6-nitro-1,3-benzodioxole-5-methanol,1,3-benzodioxole-5-methanol,6-nitro,1,3-benzodioxole-5-methanol, 6-nitro,6-nitro-1,3-benzodioxol-5-yl methanol #
519111
(6-nitro-1,3-benzodioxol-5-yl)methanol
C1OC2=C(O1)C=C(C(=C2)CO)[N+](=O)[O-]
Packaging Glass bottle
Color Yellow
Quantity 5g
Infrared Spectrum Authentic
Melting Point 121°C to 125°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 19, I, 633
Formula Weight 197.15
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 6-Nitropiperonyl alcohol, 98%
Show More Show Less