Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Nitroveratraldehyde (Tech.), 85%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
20357-25-9
C9H9NO5
211.173
MFCD00007134
YWSPWKXREVSQCA-UHFFFAOYSA-N
6-nitroveratraldehyde,dmnb,3,4-dimethoxy-6-nitrobenzaldehyde,4,5-dimethoxy-2-nitro-benzaldehyde,dna-pk inhibitor,benzaldehyde, 4,5-dimethoxy-2-nitro,2-nitro-4,5-dimethoxybenzaldehyde,dna-dependent protein kinase inhibitor,2-nitro-4,5-bis methyloxy benzaldehyde,pubchem8213
88505
4,5-dimethoxy-2-nitrobenzaldehyde
COC1=C(C=C(C(=C1)C=O)[N+](=O)[O-])OC
20357-25-9
C9H9NO5
211.173
MFCD00007134
YWSPWKXREVSQCA-UHFFFAOYSA-N
6-nitroveratraldehyde,dmnb,3,4-dimethoxy-6-nitrobenzaldehyde,4,5-dimethoxy-2-nitro-benzaldehyde,dna-pk inhibitor,benzaldehyde, 4,5-dimethoxy-2-nitro,2-nitro-4,5-dimethoxybenzaldehyde,dna-dependent protein kinase inhibitor,2-nitro-4,5-bis methyloxy benzaldehyde,pubchem8213
88505
4,5-dimethoxy-2-nitrobenzaldehyde
COC1=C(C=C(C(=C1)C=O)[N+](=O)[O-])OC
Infrared Spectrum Authentic
Quantity 25g
Color Yellow
Packaging Glass bottle
Melting Point 128°C to 135°C
CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 85%
Beilstein 08, 262
Formula Weight 211.17
Physical Form Fluffy Powder
Percent Purity ≥80%
Grade Technical
Chemical Name or Material 6-Nitroveratraldehyde, tech., 85%
Show More Show Less

AIR SENSITIVE

LIGHT SENSITIVE