Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-O-(Triisopropylsilyl)-D-galactal 97.0+%, TCI America™

Click to view available options
Quantity:
200mg
166021-01-8
C15H30O4Si
302.486
VYZHAYQIZWKLNP-RBSFLKMASA-N
1,5-Anhydro-2-deoxy-6-O-(triisopropylsilyl)-D-lyxo-hex-1-enitol
10859624
(2R,3R,4R)-2-[tri(propan-2-yl)silyloxymethyl]-3,4-dihydro-2H-pyran-3,4-diol
CC(C)[Si](C(C)C)(C(C)C)OCC1C(C(C=CO1)O)O
166021-01-8
C15H30O4Si
302.486
VYZHAYQIZWKLNP-RBSFLKMASA-N
1,5-Anhydro-2-deoxy-6-O-(triisopropylsilyl)-D-lyxo-hex-1-enitol
10859624
(2R,3R,4R)-2-[tri(propan-2-yl)silyloxymethyl]-3,4-dihydro-2H-pyran-3,4-diol
CC(C)[Si](C(C)C)(C(C)C)OCC1C(C(C=CO1)O)O
Quantity 200mg
Color Yellow
Formula Weight 302.49
Physical Form Clear Liquid at 20°C
Percent Purity ≥97.0% (GC)
Chemical Name or Material 6-O-(Triisopropylsilyl)-D-galactal