Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Phenoxy-3-pyridinamine, ≥95%, Maybridge™

Click to view available options
Quantity:
1g
10g
25g
25194-67-6
C11H10N2O
186.214
MFCD01692449
DETKIRMPBJPJRQ-UHFFFAOYSA-N
6-phenoxy-3-pyridinamine,3-amino-6-phenoxypyridine,5-amino-2-phenoxypyridine,pyridine, 5-amino-2-phenoxy,6-phenoxypyridine-3-amine,3-pyridinamine,6-phenoxy,3-pyridinamine, 6-phenoxy,6-phenoxy-pyridin-3-ylamine,6-phenoxy-3-pyridylamine,2-phenoxy-5-aminopyridine
212856
6-phenoxypyridin-3-amine
C1=CC=C(C=C1)OC2=NC=C(C=C2)N
25194-67-6
C11H10N2O
186.214
MFCD01692449
DETKIRMPBJPJRQ-UHFFFAOYSA-N
6-phenoxy-3-pyridinamine,3-amino-6-phenoxypyridine,5-amino-2-phenoxypyridine,pyridine, 5-amino-2-phenoxy,6-phenoxypyridine-3-amine,3-pyridinamine,6-phenoxy,3-pyridinamine, 6-phenoxy,6-phenoxy-pyridin-3-ylamine,6-phenoxy-3-pyridylamine,2-phenoxy-5-aminopyridine
212856
6-phenoxypyridin-3-amine
C1=CC=C(C=C1)OC2=NC=C(C=C2)N
Quantity 1g
Packaging Amber glass bottle
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 186.21
Percent Purity 95%
Chemical Name or Material 6-Phenoxy-3-pyridinamine

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.