Alfa Aesar™ 6-Phenylhexanoic acid, 98+%

Manufacturer: Alfa Aesar™ A1118706

Quantity 5g

 View more versions of this product

Catalog No. AAA1118706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5581-75-9
    Molecular Formula C6H5(CH2)5CO2H
    Formula Weight 192.26
    MDL Number MFCD00014381
    Beilstein 1950106
    Chemical Name or Material 6-Phenylhexanoic acid

    Specifications

    Formula Weight 192.26
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    CAS 5581-75-9
    MDL Number MFCD00014381
    Beilstein 1950106
    Boiling Point 157° to 160°C (0.3mmHg)
    Melting Point 17° to 19°C
    Quantity 5g
    Molecular Formula C6H5(CH2)5CO2H
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Flash Point >110°C (230°F)
    Chemical Name or Material 6-Phenylhexanoic acid
    Assay ≥98%

    Description

    Applications
    6-Phenylhexanoic acid was employed as a model compound to investigate the effects of chromophore orientation and molecular conformation on surface-enhanced Raman scattering based on metal nanostructures. It was also employed as a substrate to study the stoichiometry of side chain metabolism or complete mineralization of surrogate napthenic acids under methanogenic conditions

    Solubility
    Difficult to mix.

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit