Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Phenylnaphthalene-2-boronic acid, 98%

Click to view available options
Quantity:
250mg
1g
5g
876442-90-9
C16H13BO2
248.088
MFCD26406213
XDKCHGWZDWHBQO-UHFFFAOYSA-N
6-phenylnaphthalen-2-yl boronic acid,boronic acid, 6-phenyl-2-naphthalenyl,6-phenylnaphthalene-2-boronic acid,6-phenylphthalen-2-yl-2-boronic acid,2-phenylnaphthalen-6-yl boronic acid,6-phenyl-2-naphthalenyl boronic acid,b-6-phenyl-2-naphthalenyl boronic acid,boronic acid, 6-phenyl-2-naphthalenyl
57539613
(6-phenylnaphthalen-2-yl)boronic acid
B(C1=CC2=C(C=C1)C=C(C=C2)C3=CC=CC=C3)(O)O
876442-90-9
C16H13BO2
248.088
MFCD26406213
XDKCHGWZDWHBQO-UHFFFAOYSA-N
6-phenylnaphthalen-2-yl boronic acid,boronic acid, 6-phenyl-2-naphthalenyl,6-phenylnaphthalene-2-boronic acid,6-phenylphthalen-2-yl-2-boronic acid,2-phenylnaphthalen-6-yl boronic acid,6-phenyl-2-naphthalenyl boronic acid,b-6-phenyl-2-naphthalenyl boronic acid,boronic acid, 6-phenyl-2-naphthalenyl
57539613
(6-phenylnaphthalen-2-yl)boronic acid
B(C1=CC2=C(C=C1)C=C(C=C2)C3=CC=CC=C3)(O)O
Quantity 5g
Beilstein 11441104
Formula Weight 248.09
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 6-Phenylnaphthalene-2-boronic acid