Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Propyl-2-thiouracil, Approx. 98%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
51-52-5
C7H10N2OS
170.23
KNAHARQHSZJURB-UHFFFAOYSA-N
6-propyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one
CCCC1=CC(=O)NC(=S)N1
51-52-5
C7H10N2OS
170.23
KNAHARQHSZJURB-UHFFFAOYSA-N
6-propyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one
CCCC1=CC(=O)NC(=S)N1

P2221, 51-52-5, C7H10N2OS

Spectrum™ 6-Propyl-2-thiouracil
Packaging Amber Glass Bottle
Quantity 25 g
CAS Max % 100%
Percent Purity Approx. 98%