Learn More
Aat BioquestSupplier Diversity Partner 6-TAMRA [6-Carboxytetramethylrhodamine] *CAS 91809-67-5* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 366
6-TAMRA is the purified single isomer of 5(6)-TAMRA. It is widely used to label nucleotides and nucleic acids.
Catalog No. 50-195-2855