Learn More
Aat BioquestSupplier Diversity Partner 6-TAMRA CPG *1000 Å*  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 6051
The light-absorbing properties of TAMRA, and spectral overlap with several commonly used fluorophores - including FAM, HEX, TET and JOE, make it useful as a FRET acceptor for the dual labeled FRET probes such as Molecular Beacons.
Catalog No. 50-195-3313