Alfa Aesar™ 6-tert-Butyl-1-indanone, 97%

Manufacturer: Alfa Aesar™ H5400603

Quantity 1g

 View more versions of this product

Catalog No. AAH5400603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 162752-17-2
    Molecular Formula C13H16O
    Formula Weight 188.27
    MDL Number MFCD11590864
    Chemical Name or Material 6-tert-Butyl-1-indanone

    Specifications

    Formula Weight 188.27
    Molecular Formula C13H16O
    MDL Number MFCD11590864
    Melting Point 94°C
    Quantity 1g
    CAS 162752-17-2
    Chemical Name or Material 6-tert-Butyl-1-indanone
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit