Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-(Trifluoromethyl)indole, 97%, Thermo Scientific Chemicals

Catalog Number p-7046974
Click to view available options
Quantity:
250 mg
1 g
13544-43-9
C9H6F3N
185.149
MFCD00272316
BPYBYPREOVLFED-UHFFFAOYSA-N
6-trifluoromethyl indole, 6-trifluoromethylindole, 6-trifluoromethyl-1h-indole, 1h-indole, 6-trifluoromethyl, pubchem1713, acmc-1bscm, intermediates-zcf02044, 6-trifluoromethyl-indole, ksc910g4n
2777523
6-(trifluoromethyl)-1H-indole
C1=CC(=CC2=C1C=CN2)C(F)(F)F
13544-43-9
C9H6F3N
185.149
MFCD00272316
BPYBYPREOVLFED-UHFFFAOYSA-N
6-trifluoromethyl indole, 6-trifluoromethylindole, 6-trifluoromethyl-1h-indole, 1h-indole, 6-trifluoromethyl, pubchem1713, acmc-1bscm, intermediates-zcf02044, 6-trifluoromethyl-indole, ksc910g4n
2777523
6-(trifluoromethyl)-1H-indole
C1=CC(=CC2=C1C=CN2)C(F)(F)F

6-(Trifluoromethyl)indole is used in pharmaceutical intermediates, liquid crystal intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-(Trifluoromethyl)indole is used in pharmaceutical intermediates, liquid crystal intermediates

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point 101°C to 105°C
Quantity 250 mg
Beilstein 1242785
Solubility Information Insoluble in water.
Formula Weight 185.15
Percent Purity 97%
Chemical Name or Material 6-(Trifluoromethyl)indole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit