Alfa Aesar™ 6-(Trifluoromethyl)uracil, 97%

Manufacturer: Alfa Aesar™ 4446706

Quantity 5g

 View more versions of this product

Catalog No. AA4446706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 672-45-7
    Molecular Formula C5H3F3N2O2
    Formula Weight 180.08
    MDL Number MFCD01011762
    Chemical Name or Material 6- (Trifluoromethyl)uracil

    Specifications

    Formula Weight 180.08
    Molecular Formula C5H3F3N2O2
    Physical Form Powder
    MDL Number MFCD01011762
    CAS 672-45-7
    Quantity 5g
    Chemical Name or Material 6- (Trifluoromethyl)uracil
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit