Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Trimethylsilyl-N-tert-butyl-2,4-hexadienaldimine, ACROS Organics™

Click to view available options
Quantity:
250mg
Packaging:
Glass Bottle in Plastic Container
171814-36-1
C13H25NSi
223.435
MFCD03791051
CGIIKXSIKQLVJK-FGFCZSSHSA-N
6-trimethylsilyl-n-tert-butyl-2,4-hexadienaldimine,6-trimethylsilyl-n-tert-butyl-2,4-hexadien-1-imine,2e,4e-n-tert-butyl-6-trimethylsilylhexa-2,4-dien-1-imine
5702562
(2E,4E)-N-tert-butyl-6-trimethylsilylhexa-2,4-dien-1-imine
CC(C)(C)N=CC=CC=CC[Si](C)(C)C
171814-36-1
C13H25NSi
223.435
MFCD03791051
CGIIKXSIKQLVJK-FGFCZSSHSA-N
6-trimethylsilyl-n-tert-butyl-2,4-hexadienaldimine,6-trimethylsilyl-n-tert-butyl-2,4-hexadien-1-imine,2e,4e-n-tert-butyl-6-trimethylsilylhexa-2,4-dien-1-imine
5702562
(2E,4E)-N-tert-butyl-6-trimethylsilylhexa-2,4-dien-1-imine
CC(C)(C)N=CC=CC=CC[Si](C)(C)C
Packaging Glass Bottle in Plastic Container
Infrared Spectrum Authentic
Refractive Index 1.5120 to 1.5150 (20°C, 589nm)
NMR Complies
Color Brown-Orange to Red
Quantity 250mg
Linear Formula (CH3)3SiCH2(CH)4CHNC(CH3)3
Formula Weight 223.43
Physical Form Liquid
Chemical Name or Material 6-Trimethylsilyl-N-tert-butyl-2, 4-hexadienaldimine