Learn More
Cole-Parmer Monarch DC6-1-06-0-0-1-1-0 Recorder, 6-Channel/100-240VAC/512MB/24VDC  DFS Item
Manufacturer:  Cole-Parmer 4900624
The DataChart 6000 is like no other paperless recorder available! It has 6 or 12 universal inputs, up to 4 pulse/frequency inputs and 6 or 12 relay outputs. Networkable using standard Ethernet port; IP65/NEMA4 rated front panel. Touchscreen control.
Catalog No. 50-631-750