Learn More
ATLANTIC BIOPHARM SOLUTIONS LLC 63MM INSTA-FLOW SS INDUST PRES
Supplier:  ATLANTIC BIOPHARM SOLUTIONS LLC 2NE1/4NPTNFBTO2
Encompass_Preferred
2.5in (63mm) Insta-Flow Stainless Steel Industrial Pressure Gauge, 0 to 160 psi, 1/4in NPT, Dry Case, Bottom Mount, Polycarbonate Lens, CSE Logo, Cleaned For O2 Service.
Catalog No. NC1758772