Learn More
SCS 6800 Series 2-Layer Rubber Mat
Provides a worksurface that does not generate a static charge
Supplier:  SCS 6840
 View more versions of this product
Catalog No. 18-688-262
Add to cart
Description
Description
  • Dissipative Dual Layer Rubber Material
    Provides a worksurface that does not generate a static charge and will control the discharge rate from all conductors (including ESD susceptible items) that are placed on the surface.
  • Dissipative (1 x 106 to <1 x 109 ohms Rtt) Top Color Layer
    Meets ANSI/ESD S20.20 worksurface required limit and recommendation of ANSI/ESD S4.1 for contact with ESD susceptible items.
  • Conductive (<1 x 104 Rtt) Black Bottom Layer
    Reduces the resistance to ground (Rtg) from the top of the surface to the groundable point ground and allows the mats to be used with ESD workstation monitors.
  • High Strength Nitrile Rubber Compound
    Designed to withstand abrasion, stretching, and tearing and may be used in soldering applications with flux and other chemicals.
Specifications
Specifications
0.072 in.
15.24 m
Gray
Meets ANSI/ESD S20.20 required limit tested per ANSI/ESD S4.1 and ESD TR53
60.96 cm
50 ft.
50 ft. x 24 in.
Rubber
24 in.