Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 6877002

CD40-TRAF6 interaction inhibitor

$92.70 - $389.00

Specifications

Chemical Name or Material 3-[(2,5-Dimethylphenyl)amino]-1-phenyl-2-propen-1-one
Molecular Formula C17H17NO
CAS 433249-94-6
Product Type Target: Additional Cytokine Receptor Products
Formula Weight 251.32
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
560210 View Documents Tocris Bioscience™
5602/10
10mg Each for $92.70
Add to cart
 
560250 View Documents Tocris Bioscience™
5602/50
50mg Each for $389.00
Add to cart
 
Description

Description

CD40-TRAF6 interaction inhibitor. Suppresses CD40-induced IL-1β and IL-6 expression in macrophages in vitro. Improves insulin sensitivity, reduces adipose tissue inflammation, and ameliorates hepatosteatosis in diet-induced obese mice.
Specifications

Specifications

3-[(2,5-Dimethylphenyl)amino]-1-phenyl-2-propen-1-one
433249-94-6
251.32
C17H17NO
Target: Additional Cytokine Receptor Products
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit