Learn More
BrandTech™ BRAND™ BIO-CERT™ Filter Tips, 0.5 to 10uL Available on GSA/VA Contract for Federal Government customers only.
Manufactured from high-purity, oleamide- and diHEMDA-free polypropylene. BrandTech™ BRAND™ BIO-CERT™ Filter Tips, 0.5 to 10µL are sterile and free of biological contaminants. Non self-sealing PE filters allow sample recovery with wetted filters. Slender design allows pipetting without touching microtube and microplate walls. Graduations provided at 2 and 10µL for quick volume check.
Manufacturer:  BrandTech Scientific 732824
 View more versions of this product
Catalog No. 13-889-203
  Check Availability
Add to cart
Specifications
Description
  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Independently certified sterile and free of biological contaminants
  • Non self-sealing PE filters allow sample recovery with wetted filters
  • Slender design
  • Thin-walled for high translucency, easy ejection, and low waste
  • Graduations at 2 and 10µL for quick volume check
  • Available in either standard or ultra low retention types
  • Available in convenient, ecological rack format, racked tips are colorless and placed into gray-colored tip tray
Ultra low retention tips
  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface for low surface tension and low sample retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain proteins, nucleic acids, detergents and other difficult liquids that can cause sample retention
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention or filter properties
Description & Specifications
Specifications
Filter Tip
0.5 to 10μL
Colorless
Yes
Polypropylene
960 Pack
4.6cm
High translucency, easy ejection and low waste, quick volume check
Sterile
2, 10μL
Ultra Low Retention, Nano-cap
TipBox, 10 boxes of 96 pieces
1.81 in.
DNA, RNase, ATP and endotoxin-free