BrandTech™ BRAND™ BIO-CERT™ Filter Tips, 0.5 to 10uL Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: BrandTech Scientific 732624

Catalog No. 13-889-165

  Check Availability
Add to Cart

    Specifications

    Graduations 2, 10μL
    Tip Style Standard
    Packaging TipRefill, 10 racks of 96 pieces
    Quantity 960 Pack
    Certifications/Compliance DNA, RNase, ATP and endotoxin-free
    For Use With (Application) High translucency, easy ejection and low waste, quick volume check
    Volume (Metric) 0.5 to 10μL
    Sterility Yes
    Color Colorless

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit