Learn More
Hanna Pharmaceutical Supply Co., Inc. SUTURE, SILK BLK BR 7-0 18"" TG140-8 1
Supplier:  Hanna Pharmaceutical Supply Co., Inc. 7733G
Encompass_Preferred
Suture with Needle Perma-Hand™ Nonabsorbable Coated Black Suture Braided Silk Size 7 - 0 18 Inch Suture Double-Armed 6.5 mm Length 3/8 Circle Micropoint - Spatula Needle 12/BX
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
Catalog No. NC0304173
Due to product restrictions, please to purchase or view availability for this product.