Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-(2,3-Dihydroxypropyl)theophylline 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
479-18-5
C10H14N4O4
254.24
MFCD00005759
KSCFJBIXMNOVSH-UHFFFAOYSA-N
dyphylline,diprophylline,diphylline,7-2,3-dihydroxypropyl theophylline,diprophyllin,glyphylline,neothylline,lufyllin,aristophyllin,diprofilline
3182
CHEBI:4728
7-(2,3-dihydroxypropyl)-1,3-dimethylpurine-2,6-dione
CN1C2=C(C(=O)N(C1=O)C)N(C=N2)CC(CO)O
479-18-5
C10H14N4O4
254.24
MFCD00005759
KSCFJBIXMNOVSH-UHFFFAOYSA-N
dyphylline,diprophylline,diphylline,7-2,3-dihydroxypropyl theophylline,diprophyllin,glyphylline,neothylline,lufyllin,aristophyllin,diprofilline
3182
CHEBI:4728
7-(2,3-dihydroxypropyl)-1,3-dimethylpurine-2,6-dione
CN1C2=C(C(=O)N(C1=O)C)N(C=N2)CC(CO)O
Loss on Drying 0.5% max. (1 g, 105°C)
Quantity 100g
Color White
Infrared Spectrum Authentic
Melting Point 160°C to 165°C
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Merck Index 15, 3526
Solubility Information Solubility in water: 1g/3mL (25 c). Other solubilities: 2g/100mL ethanol,1g/100mL chloroform
Formula Weight 254.24
Physical Form Powder
Percent Purity ≥98.5% (on dried substance)
Chemical Name or Material 7-(2, 3-Dihydroxypropyl)theophylline, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 207-526-1

RTECSNumber : XH5100000

TSCA : TSCA