Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Amino-2-methylindazole, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
90223-02-2
C8H9N3
147.18
KNZJAZZOKTUEHM-UHFFFAOYSA-N
7-amino-2-methylindazole,2-methyl-2h-indazol-7-amine,7-amino-2-methyl-2h-indazole,2-methyl-2h-indazol-7-ylamine,2h-indazol-7-amine,2-methyl
1480773
2-methylindazol-7-amine
CN1C=C2C=CC=C(C2=N1)N
90223-02-2
C8H9N3
147.18
KNZJAZZOKTUEHM-UHFFFAOYSA-N
7-amino-2-methylindazole,2-methyl-2h-indazol-7-amine,7-amino-2-methyl-2h-indazole,2-methyl-2h-indazol-7-ylamine,2h-indazol-7-amine,2-methyl
1480773
2-methylindazol-7-amine
CN1C=C2C=CC=C(C2=N1)N
Melting Point 65.9°C to 66.4°C
Packaging Glass Bottle
Color White to Brown
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 147.18
Physical Form Powder or Crystalline Powder
Percent Purity 97%
Chemical Name or Material 7-Amino-2-methylindazole
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning