Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Aminocephalosporanic acid, 95-102%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
957-68-6
C10H12N2O5S
272.27
MFCD00005177
HSHGZXNAXBPPDL-HZGVNTEJSA-N
7-aminocephalosporanic acid, 7-aca, 7-aminocephalosporinic acid, 7beta-aminocephalosporanic acid, 7-acs, unii-9xi67897rg, 7r-7-aminocephalosporanic acid, 7r-7-aminocephalosporanate, cephalosporanic acid, 7-amino, 6r,7r-3-acetoxymethyl-7-amino-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid
441328
CHEBI:2255
(6R,7R)-3-(acetyloxymethyl)-7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
CC(=O)OCC1=C(N2C(C(C2=O)N)SC1)C(=O)O
957-68-6
C10H12N2O5S
272.27
MFCD00005177
HSHGZXNAXBPPDL-HZGVNTEJSA-N
7-aminocephalosporanic acid, 7-aca, 7-aminocephalosporinic acid, 7beta-aminocephalosporanic acid, 7-acs, unii-9xi67897rg, 7r-7-aminocephalosporanic acid, 7r-7-aminocephalosporanate, cephalosporanic acid, 7-amino, 6r,7r-3-acetoxymethyl-7-amino-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid
441328
CHEBI:2255
(6R,7R)-3-(acetyloxymethyl)-7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
CC(=O)OCC1=C(N2C(C(C2=O)N)SC1)C(=O)O
Melting Point >300°C
Specific Rotation + 94
Quantity 1g
Packaging Glass bottle
Color White to Beige
Infrared Spectrum Authentic
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (HPLC)
Merck Index 15, 429
Solubility Information Solubility in water: soluble
Formula Weight 272.27
Physical Form Crystalline Powder
Percent Purity 95-102%
Chemical Name or Material 7-Aminocephalosporanic acid, 95-102%
Show More Show Less

GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

GHS P Statement
Wear eye protection/face protection.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor

GHS Signal Word: Danger

EINECSNumber : 213-485-0